Merck
  • Toxic effects of aminophenols on aquatic life using the zebrafish embryo test and the comet assay.
Toxic effects of aminophenols on aquatic life using the zebrafish embryo test and the comet assay.
Bulletin of environmental contamination and toxicology (2004-09-25)
L W Sun, M M Qu, Y Q Li, Y L Wu, Y G Chen, Z M Kong, Z T Liu
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Aminophenol, 98%
Supelco
3-Aminophenol, PESTANAL®, analytical standard