- Development of a simple system for dehydrocondensation using solid-phase adsorption of a water-soluble dehydrocondensing reagent (DMT-MM).
Development of a simple system for dehydrocondensation using solid-phase adsorption of a water-soluble dehydrocondensing reagent (DMT-MM).
Chemical & pharmaceutical bulletin (2004-10-07)
Yasunobu Watanabe, Takako Fuji, Kazuhito Hioki, Shohei Tani, Munetaka Kunishima
PMID15467240
ZUSAMMENFASSUNG
It has been indicated that hydrophilic solid powder to which aqueous solution of a novel dehydrocondensing reagent DMT-MM is adsorbed becomes a simple solid-phase dehydrocondensing reagent of low cost. Reaction in a liquid--liquid biphasic system on the surface of a solid phase with a large area was accelerated by suspending this powder in a dichloromethane solution of a carboxylic acid and an amine to be condensed. The reaction was rapid with a high yield despite the heterogeneity of the system. Like general solid-phase reagents, a hydrophobic carboxamide alone could be isolated at a relatively high purity only by filtration of the resulting suspension of reaction mixture.