Merck
Detection of bacteria with carbohydrate-functionalized fluorescent polymers.
Journal of the American Chemical Society (2004-10-14)
Matthew D Disney, Juan Zheng, Timothy M Swager, Peter H Seeberger
ZUSAMMENFASSUNG
Many pathogens that infect humans use cell surface carbohydrates as receptors to facilitate cell-cell adhesion. The hallmark of these interactions is their multivalency, or the simultaneous occurrence of multiple interactions. We have used a carbohydrate-functionalized fluorescent polymer, which displays many carbohydrate ligands on a single polymer chain, to allow for multivalent detection of pathogens. Incubation of a mannose-functionalized polymer with Escherichia coli yields brightly fluorescent aggregates of bacteria. These results show that carbohydrate-functionalized fluorescent polymers are a versatile detection method for bacteria. Future design of detectors for other pathogens only requires information on the carbohydrates bound by the organisms, which has been exhaustively reported in the literature.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Diisopropylethylamin, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Diisopropylethylamin, 99.5%, biotech. grade
Sigma-Aldrich
N,N-Diisopropylethylamin, purified by redistillation, 99.5%
Sigma-Aldrich
N-Ethyldiisopropylamin, ≥98.0%
Sigma-Aldrich
N-Ethyldiisopropylamin -Lösung, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone