Merck
[Selective nutrient medium for the isolation of Bacillus cereus].
Zhurnal mikrobiologii, epidemiologii, i immunobiologii (2004-10-16)
Z Z Sultanov, L S Kulakova, L G Perepelitsa, S K Abdulganieva
ZUSAMMENFASSUNG
A selective dried medium for the isolation of B. cereus from clinical material and foodstuffs has been developed. The medium has high selective properties which ensure the isolation of B. cereus from microbial association in pure culture, thus making it possible to accelerate further identification of the microorganisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithiumsulfat, ≥98.5% (titration)
Sigma-Aldrich
Lithiumsulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Lithiumsulfat, purum p.a., ≥98.0% (T)