- Coupled-enzyme system for the determination of lipase activity.
Coupled-enzyme system for the determination of lipase activity.
Biotechnology letters (2004-10-16)
Ines Ben Rejeb, Jeannette Ben Hamida, Mohamed Gargouri
PMID15483386
ZUSAMMENFASSUNG
A new method for lipase activity determination is described, in which liberation of fatty acids by lipase-catalyzed hydrolysis is coupled to lipoxygenation. The second reaction forms hydroperoxy-fatty acids containing a conjugated double bond that are detected at 234 nm. The method is sensitive, cheap and easy to use when compared to a titration method.
MATERIALIEN