Merck
The reactivity of 1,3-butadiene with butadiene-derived popcorn polymer.
Journal of hazardous materials (2004-11-03)
M E Levin, A D Hill, L W Zimmerman, T E Paxson
ZUSAMMENFASSUNG
Adiabatic calorimetry performed on butadiene-derived popcorn polymer samples from industrial facilities has revealed exothermic behavior accompanied by non-condensible gas production, indicative of possible decomposition, at elevated temperatures. In the presence of low concentrations of 1,3-butadiene, reactivity is observed at temperatures of 60-70 degrees C; that is, 20-30 degrees C below those usually seen for butadiene alone. Once the butadiene is consumed, the reaction behavior reverts to that of the popcorn polymer alone. At higher butadiene concentrations, the low temperature reaction persists, eventually merging with typical butadiene behavior. The butadiene reactivity with popcorn polymer is attributed to polymerization reaction at free radical sites in the popcorn polymer. Different popcorn polymer samples exhibit distinct extents of reactivity, presumably depending on the nature and concentration of the free radical sites and the structure of the material. Uninhibited butadiene exposed to 100 psia air, which may act to generate peroxide species, shows a small, additional exotherm around 50-80 degrees C. Contact of butadiene with lauroyl peroxide, providing free radicals upon decomposition, generates an exotherm at temperatures as low as 60 degrees C.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Luperox® LP, Lauroylperoxid, ≥98%