Merck
  • [Studies on the analgesic and anti-inflammatory effect of bornyl acetate in volatile oil from Amomum villosum].
[Studies on the analgesic and anti-inflammatory effect of bornyl acetate in volatile oil from Amomum villosum].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2004-11-05)
Xiaosong Wu, Xiaoguang Li, Fei Xiao, Zhidong Zhang, Zhenxia Xu, Huan Wang
ZUSAMMENFASSUNG
To study analgesic and anti-inflammatory effects of bornyl acetate, the main ingredient of Amomum villosum volatile oil. The analgesic effects were tested by hot-plate and writhing reaction method, the ear swelling caused by dimethylbenzene in mice. Bornyl acetate could restrain writhing reaction caused by acetic acid glacial, lighten the pain caused by hot-plate. It could also suppress ear swelling caused by dimethylbenzene in mice. Bornyl acetate shows analgesic and anti-inflammatory effects.