Merck
  • Determination of manganese in some medicinal plants and their infusions by a catalytic spectrophotometric method.
Determination of manganese in some medicinal plants and their infusions by a catalytic spectrophotometric method.
Annali di chimica (2005-01-01)
Konstantin Mutaftchiev
ZUSAMMENFASSUNG
The catalytic effect of manganese (II) on the oxidation of the azo dye 3-Methyl-6-(2-hydroxyethoxy)-2-[2-methoxy-4-N (N, N diethylamino) phenylazo] benzothiazolium methylsulphate, with potassium periodate in the presence of 1,10-phenanthroline in weakly acidic media was studied. The reaction was followed spectrophotometrically by measuring the decrease in the absorbance of the dye at 560 nm. Under the optimum conditions (4 x 10(-5) mol dm(-3) azo dye, 4 x 10(-4) mol dm(-3) potassium periodate, 1 x 10(-4) mol dm(-3) 1,10-phenanthroline, 0.1 mol dm(-3) buffer--pH 3.0, 70 degrees C, 8 min) manganese (II) in the range 0.1-5 ng cm(-3) could be determined by the fixed-time method with a detection limit of 0.035 ng cm(-3). The developed method is highly sensitive, selective, and simple. The method was applied successfully to the determination of total manganese in some medicinal plants and to analyse their infusions for trace amounts of total manganese and free manganese (II) ions without separation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kalium(meta)periodat, 99.8%, ACS reagent