Merck
  • Incorporating targeted treatments into standard regimens: whither the role of established doses?
Incorporating targeted treatments into standard regimens: whither the role of established doses?
Cancer investigation (2005-01-12)
Don S Dizon
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
CI−994, ≥98% (HPLC), powder