Merck
  • [Quantitative determination of ethyl-p-hydroxybenzoate in resins extracted from Dracaena cochinchinensis with two technologies].
[Quantitative determination of ethyl-p-hydroxybenzoate in resins extracted from Dracaena cochinchinensis with two technologies].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2005-02-15)
Yue-ying Song, Qing-yun Zhang, Ying-qing Hu, Chang-hu Deng
ZUSAMMENFASSUNG
Quantitative determination was made of ethyl-p-hydroxybenzoate in Dracaena cochinchinensis extracted with two technologies. The sample was resolved with methanol and isolated by TLC, purged with methanol. Tge sample solution was chroma to graphed on a C18 column with acetonitrile-1% acetic acid (31:69) as mobile phase, detecting at 257 nm and content was calculated with external standard method. The standard curves of ethyl-p-hydroxybenzoate were linear in the range of 0.206-4.12 ng, r = 0.9998. The average recovery was 97.2% and RSD was 1.4%. The content of ethyl-p-hydroxybenzoate in D. cochinchinensis extracted with heating-tree technongy is higher than that with traditional technology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Hydroxy-benzoesäure-ethylester, ReagentPlus®, 99%
Supelco
Ethylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Ethylparahydroxybenzoat, European Pharmacopoeia (EP) Reference Standard