Merck
  • Determination of 14 benzodiazepines and hydroxy metabolites, zaleplon and zolpidem as tert-butyldimethylsilyl derivatives compared with other common silylating reagents in whole blood by gas chromatography-mass spectrometry.
Determination of 14 benzodiazepines and hydroxy metabolites, zaleplon and zolpidem as tert-butyldimethylsilyl derivatives compared with other common silylating reagents in whole blood by gas chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-03-01)
Teemu Gunnar, Kari Ariniemi, Pirjo Lillsunde
ZUSAMMENFASSUNG
The most common commercially available silylating reagents, N-methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA), N,O-bis-(trimethylsilyl)trifluoroacetamide+1% trimethylchlorosilane (BSTFA+1% TMCS) and N-methyl-N-(tert-butyldimethylsilyl)trifluoroacetamide (MTBSTFA) were evaluated to achieve optimal derivatization conditions for analyzing various benzodiazepines based on gas chromatography-electron impact ionization-mass spectrometry (GC-EI-MS). Sensitivity, repeatability, retention times and stability of the derivatives, as well as specificity of mass fragmentation, were studied in detail. Also other parameters affecting the derivatization chemistry of benzodiazepines are discussed. tert-Butyldimethylsilyl (TBDMS) derivatives proved to be more stable, reproducible and sensitive than corresponding trimethylsilyl (TMS) derivatives for the GC-EI-MS analysis of benzodiazepines. Based on the TBDMS derivatives, a rapid, reliable, sensitive and quantitative GC-MS method was developed for the determination of 14 benzodiazepines and two hydroxy metabolites, as well as two non-benzodiazepine hypnotic agents, zolpidem and zaleplon, using 50 microl of whole blood. The method was completely validated in terms of accuracy, intra- and interday precision, limit of detection (LOD), limit of quantitation (LOQ), linearity, selectivity and extraction efficiency; these were all within the required limits, except for the accuracy of nitrazepam at a medium concentration level.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
N-Methyl-N-trimethylsilyltrifluoracetamid, derivatization grade (GC derivatization), LiChropur, ≥98.5%
Supelco
N-Methyl-N-trimethylsilyltrifluoracetamid, synthesis grade
Supelco
N-Methyl-N-trimethylsilyltrifluoracetamid, BioReagent, for silylations, LiChropur
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoracetamid, >97%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoracetamid mit 1% tert-Butyldimethylchlorsilan, ≥95%
Supelco
N-Methyl-N-trimethylsilyl-trifluoracetamid mit 1% Trimethylchlorsilan, derivatization grade (GC derivatization), LiChropur
Supelco
N-tert-Butyldimethylsilyl-N-methyltrifluoracetamid mit 1% tert-Butyldimethylchlorsilan, ampule of 5 × 1 mL, (with 1% t-BDMCS), analytical standard, Cerilliant®
Supelco
N-Methyl-N-trimethylsilyltrifluoracetamid, ampule of 10 × 1.2 mL, analytical standard, Cerilliant®