Merck
  • Kinetic spectrophotometric determination of rutin by its inhibitory effect on the oxidation of amaranth by potassium periodate.
Kinetic spectrophotometric determination of rutin by its inhibitory effect on the oxidation of amaranth by potassium periodate.
Annali di chimica (2005-04-02)
Shuhao Wang, Lingyun Du, Xingjun Yao, Xueli Niu, Huisheng Zhuang
ZUSAMMENFASSUNG
A novel kinetic spectrophotometric method is described for the determination of rutin. The method is based on the inhibitory effect of rutin on the oxidation reaction of amaranth by potassium periodate in acidic media at 100 degrees C. The linear range for the determination of rutin is 0.02 - 0.50 microg/ml, and the detection limit is 0.014 microg/ml. The method has been successfully applied to the determination of rutin in medicine of rutin tablet and traditional Chinese medicine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kalium(meta)periodat, 99.8%, ACS reagent