Merck
  • 4-acetoxyscirpendiol of Paecilomyces tenuipes inhibits Na(+)/D-glucose cotransporter expressed in Xenopus laevis oocytes.
4-acetoxyscirpendiol of Paecilomyces tenuipes inhibits Na(+)/D-glucose cotransporter expressed in Xenopus laevis oocytes.
Journal of biochemistry and molecular biology (2005-04-14)
Ocki Yoo, Joo-Hiuk Son, Dong-Hee Lee
ZUSAMMENFASSUNG
Cordyceps, an entomopathogenic fungus, contains many health-promoting ingredients. Recent reports indicate that the consumption of cordyceps helps reduce blood-sugar content in diabetics. However, the mechanism underlying this reduction in circulatory sugar content is not fully understood. Methanolic extracts were prepared from the fruiting bodies of Paecilomyces tenuipes, and 4-beta acetoxyscirpendiol (4-ASD) was eventually isolated and purified. Na(+)/Glucose transporter-1 (SGLT-1) was expressed in Xenopus oocytes, and the effect of 4-ASD on SGLT-1 was analyzed utilizing a voltage clamp and by performing 2-deoxy-D-glucose (2-DOG) uptake studies. 4-ASD was shown to significantly inhibit SGLT-1 activity compared to the non-treated control in a dose-dependent manner. In the presence of the derivatives of 4-ASD (diacetoxyscirpenol or 15-acetoxyscirpendiol), SGLT-1 activity was greatly inhibited in an 4-ASD-like manner. Of these derivatives, 15-acetoxyscirepenol inhibited SGLT-1 as well as 4-ASD, whereas diacetoxyscirpenol was slightly less effective. Taken together, these results strongly indicate that 4-ASD in P. tenuipes may lower blood sugar levels in the circulatory system. We conclude that 4-ASD and its derivatives are effective SGLT-1 inhibitors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Diacetoxyscirpenol -Lösung, ~100 μg/mL in acetonitrile, analytical standard