Merck
  • Induction of hepatic peroxisome proliferation by 8-2 telomer alcohol feeding in mice: formation of perfluorooctanoic acid in the liver.
Induction of hepatic peroxisome proliferation by 8-2 telomer alcohol feeding in mice: formation of perfluorooctanoic acid in the liver.
Toxicological sciences : an official journal of the Society of Toxicology (2005-05-13)
Naomi Kudo, Yuko Iwase, Hiroshi Okayachi, Yoshihiro Yamakawa, Yoichi Kawashima
ZUSAMMENFASSUNG
The effects of dietary administration of 1H, 1H, 2H, 2H-perfluorodecanol (8-2 telomer alcohol), on peroxisome proliferation in the liver of mice were studied. Male ddY mice were fed on a diet containing 8-2 telomer alcohol at concentrations of 0, 0.025, 0.05, 0.1, and 0.2% (w/w) for 7, 14, 21, and 28 days. These treatments with 8-2 telomer alcohol caused liver enlargement in a dose- and duration-dependent manner. Peroxisome proliferation in the liver of mice was confirmed by electron microscopic examination. Peroxisomal acyl-CoA oxidase was induced by these treatments with 8-2 telomer alcohol in a dose- and time-dependent manner. The concentration of perfluorooctanoic acid (PFOA) and related compounds were determined in the liver and plasma, since PFOA had been shown to be a possible metabolite of 8-2 telomer alcohol and to cause significant peroxisome proliferation in rodents. Five metabolites, namely, perfluorooctanoic acid (PFOA), perfluorononanoic acid (PFNA), 2H, 2H-perfluorodecanoic acid (8-2 telomer acid), and two unidentified metabolites, were present in the liver and serum. PFOA was confirmed to be accumulated in the liver of mice following the administration of 8-2 telomer alcohol in a dose- and duration-dependent manner. A linear relationship was observed between the concentration of PFOA and the activity of peroxisomal acyl-CoA oxidase in the liver of mice. These results strongly suggest that PFOA, but not 8-2 telomer alcohol itself, caused peroxisome proliferation in the liver. The present study provided evidence that 8-2 telomer alcohol is converted into PFOA in vivo and that the PFOA formed produces biological effects in the liver of mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1H,1H,2H,2H-Perfluor-1-decanol, 97%