Merck
  • Adsorptive stripping voltammetric technique for the rapid determination of tobramycin on the hanging mercury electrode.
Adsorptive stripping voltammetric technique for the rapid determination of tobramycin on the hanging mercury electrode.
Journal of pharmaceutical and biomedical analysis (2005-06-01)
Nan Sun, Wei-Min Mo, Zhen-Lu Shen, Bao-Xiang Hu
ZUSAMMENFASSUNG
A linear sweep adsorptive stripping voltammetric method (AdS-LSV) for the determination of tobramycin (TOB) has been proposed for the first time. The method is based on the formation of the voltammetrically active iso-butyraldehyde derivative of TOB and the electrochemical behavior of TOB iso-butyraldehyde derivative has been studied. TOB iso-butyraldehyde derivative exhibits a sensitive cathodic peak at -1.40 V (versus SCE) in a medium of B-R buffer (pH 9.8) with a scan rate of 90 mVs(-1) after a preconcentration period of 120 s at -1.10 V (versus SCE). The linear concentration range of application was 6.87 x 10(-9) - 3.44 x 10(-7) mol L(-1) of TOB, with a relative standard deviation of 4.4% (for a level of 1.0 x 10(-7) mol L(-1)) and a detection limit of 3.44 x 10(-9) mol L(-1). The method was applied to the direct determination of TOB in injectable formulations and spiked urine and serum samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Butyraldehyd, purified by redistillation, ≥99.5%
Sigma-Aldrich
Butyraldehyd, ≥96.0%
Sigma-Aldrich
Butyraldehyd, natural, FG