Merck
  • Preparation of microencapsulated phase-change materials (MCPCMs) by means of interfacial polycondensation.
Preparation of microencapsulated phase-change materials (MCPCMs) by means of interfacial polycondensation.
Journal of microencapsulation (2005-07-16)
Y-H Tseng, M-H Fang, P-S Tsai, Y-M Yang
ZUSAMMENFASSUNG
Microcapsules for energy storage and/or heat transfer applications containing phase-change materials (PCMs-including n-pentadecane, n-eicosane and a paraffin wax) were successfully produced by emulsifying the PCMs as small droplets in an aqueous, water-soluble urea-formaldehyde pre-polymer solution substantially free of emulsifier while polymerizing the pre-polymer at the interface by acid-catalyst. The core/shell structured microcapsules were also characterized with size distribution analysis, scanning electron microscopy, FTIR spectrometry and differential scanning calorimetry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eicosan, 99%
Sigma-Aldrich
Pentadecan, ≥99%
Sigma-Aldrich
Pentadecan, ≥98.0% (GC)
Supelco
Pentadecan, analytical standard
Supelco
Eicosan, analytical standard