Merck
[Studies on the chemical constituents in seeds of Atriplex centralasiatica].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2005-08-04)
Yang Zhang, Yi-min Zhao
ZUSAMMENFASSUNG
To study the chemical constituents in seeds of Atriplex centralasiatica. The chemical components were isolated by solvents extractions and column chromatography. The chemical structures were identified on the basis of physicochemical constants and spectral data. Six compounds were isolated and identified as isorhamnetin(I), tricin(II), querecetin-7-O-alpha-L-rhamnoside(III), isoorientin(IV), beta-sitosterol(V), beta-daucosterlo(VI). Compound I, II, III, IV are isolated from genus Atriplex for the first time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isoorientin, ≥98% (HPLC)
Isoorientin, primary reference standard