Merck
  • Separation and detection of the isomeric equine conjugated estrogens, equilin sulfate and delta8,9-dehydroestrone sulfate, by liquid chromatography--electrospray-mass spectrometry using carbon-coated zirconia and porous graphitic carbon stationary phases.
Separation and detection of the isomeric equine conjugated estrogens, equilin sulfate and delta8,9-dehydroestrone sulfate, by liquid chromatography--electrospray-mass spectrometry using carbon-coated zirconia and porous graphitic carbon stationary phases.
Journal of chromatography. A (2005-08-05)
John C Reepmeyer, James F Brower, Hongping Ye
ZUSAMMENFASSUNG
Equilin-3-sulfate and delta8,9-dehydroestrone-3-sulfate are two isomers found in equine conjugated estrogens that differ in structure only by the position of a double bond in the steroid B-ring. These geometric isomers were not resolved on a C18 column during the analysis of conjugated estrogen drug products by LC-MS using acetonitrile-ammonium acetate buffer as the mobile phase. While no separations of these two isomers were observed on C18 or other alkyl-bonded silica based phases using a variety of mobile phase conditions, partial separations were achieved on phenyl bonded silica phases with a resolution of 1.5 on a diphenyl phase, and baseline separations were readily achieved on two carbonaceous phases with resolutions routinely exceeding three on graphitic carbon-coated zirconia (Zr-CARB) and resolutions as high as 19 on porous graphitic carbon (Hypercarb). An examination of a selected few conjugated estrogens in the complex drug substance by LC-MS on Hypercarb is presented.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Equilin