Merck
[Three cases of occupational chronic allyl chloride poisoning].
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases (2005-08-30)
Jun Meng, Li Guo, Feng-jie Jiang, Gui-fang Li
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Allylchlorid, reagent grade, 98%
Sigma-Aldrich
Allylchlorid, ReagentPlus®, 99%
SAFC
Allylchlorid