Merck
  • Effect of different decoquinate treatments on cryptosporidiosis in naturally infected cashmere goat kids.
Effect of different decoquinate treatments on cryptosporidiosis in naturally infected cashmere goat kids.
The Veterinary record (2005-08-30)
I Ferre, A Benito-Peña, U García, K Osoro, L M Ortega-Mora
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Decochinat, VETRANAL®, analytical standard