Merck
  • Separation of some chiral flavonoids by microbial cyclosophoraoses and their sulfated derivatives in micellar electrokinetic chromatography.
Separation of some chiral flavonoids by microbial cyclosophoraoses and their sulfated derivatives in micellar electrokinetic chromatography.
Electrophoresis (2005-09-17)
Heylin Park, Seunho Jung
ZUSAMMENFASSUNG
Neutral cyclosophoraoses (Cys) and highly sulfated cyclosophoraoses (HS-Cys) were successfully applied as chiral selectors with SDS for the separation of some chiral flavonoids in MEKC. HS-Cys were synthesized by the chemical modification of a family of neutral Cys isolated from a soil microorganism, Rhizobium meliloti 2011. Chiral catechin was separated with a resolution (R(s)) of 0.754 by neutral Cys and SDS. In the case of isosakuranetin and neohesperidin, resolution (R(s)) values of 1.483 and 1.306 were obtained with HS-Cys and SDS, respectively.