Merck
  • Microscale sample deposition onto hydrophobic target plates for trace level detection of neuropeptides in brain tissue by MALDI-MS.
Microscale sample deposition onto hydrophobic target plates for trace level detection of neuropeptides in brain tissue by MALDI-MS.
Journal of mass spectrometry : JMS (2005-10-12)
Hui Wei, Stacey L Dean, Mark C Parkin, Kerstin Nolkrantz, James P O'Callaghan, Robert T Kennedy
ZUSAMMENFASSUNG
A sample preparation method that combines a modified target plate with a nanoscale reversed-phase column (nanocolumn) was developed for detection of neuropeptides by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS). A gold-coated MALDI plate was modified with an octadecanethiol (ODT) self-assembled monolayer to create a hydrophobic surface that could concentrate peptide samples into a approximately 200-500-microm diameter spot. The spot sizes generated were comparable to those obtained for a substrate patterned with 200-microm hydrophilic spots on a hydrophobic substrate. The sample spots on the ODT-coated plate were 100-fold smaller than those formed on an unmodified gold plate with a 1-microl sample and generated 10 to 50 times higher mass sensitivity for peptide standards by MALDI-TOF MS. When the sample was deposited on an ODT-modified plate from a nanocolumn, the detection limit for peptides was as low as 20 pM for 5-microl samples corresponding to 80 amol deposited. This technique was used to analyze extracts of microwave-fixed tissue from rat brain striatum. Ninety-eight putative peptides were detected including several that had masses matching neuropeptides expected in this brain region such as substance P, rimorphin, and neurotensin. Twenty-three peptides had masses that matched peaks detected by capillary liquid chromatography with electrospray ionization MS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Octadecanethiol, 98%
Sigma-Aldrich
1-Octadecanethiol, purum, ≥95.0% (GC)