Merck
  • N-acetylcysteine and S-methylcysteine inhibit MeIQx rat hepatocarcinogenesis in the post-initiation stage.
N-acetylcysteine and S-methylcysteine inhibit MeIQx rat hepatocarcinogenesis in the post-initiation stage.
Carcinogenesis (2005-12-13)
Motome Nishikawa-Ogawa, Hideki Wanibuchi, Keiichirou Morimura, Anna Kinoshita, Takayuki Nishikawa, Shuji Hayashi, Yoshihisa Yano, Shoji Fukushima
ZUSAMMENFASSUNG
N-acetylcysteine (NAC) and S-methylcysteine (SMC), water soluble organosulfur compounds contained in garlic, were evaluated for chemoprevention of hepatocarcinogenesis after 2-amino-3,8-dimethylimidazo[4,5-f]quinoxaline (MeIQx) initiation in rats. Intergastric treatment with NAC or SMC five times a week resulted in decreased numbers and areas of preneoplastic, glutathione S-transferase placental form (GST-P) positive foci of the liver in a dose-dependent manner. Moreover, cell proliferation was reduced in GST-P positive foci by NAC and SMC. Insulin-like growth factor I (IGF-I) and inducible nitric oxide synthase (iNOS) mRNA expressions were found downregulated in the liver by NAC. The studies indicate that NAC can serve as a chemopreventive agent for rat hepatocarcinogenesis induced by MeIQx by reducing cell proliferation, which may involve IGF-I and iNOS downregulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
S-Methyl-L-Cystein, substrate for methionine sulfoxide reductase A