Merck
Allyl chloride-induced time dependent changes of lipid peroxidation in rat nerve tissue.
Neurochemical research (2005-12-13)
Qing-Shan Wang, Ke-Qin Xie, Cui-li Zhang, Ying-Jian Zhu, Li-Ping Zhang, Xin Guo, Su-fang Yu
ZUSAMMENFASSUNG
To accurately know the time-dependent changes of the lipid peroxidation and antioxidative status for elucidating the mechanism of neuropathy induced by allyl chloride (AC), the malondialdehyde (MDA), anti-reactive oxygen species (anti-ROS), glutathione (GSH), catalase (CAT), glutathione peroxidase (GPx) and superoxide dismutase (SOD) were investigated in cerebrum, spinal cord and sciatic nerve of rats after 0, 3, 6, 9, 12 weeks of AC administration. AC was administrated to Wistar rats by gavage at a single dosage of 200 mg/kg/per dose (three times per week). Rats were sacrificed after 0, 3, 6, 9, 12 weeks of treatment, and cerebrum, spinal cord, sciatic nerves were dissected, homogenized and used for the determination of lipid peroxidation and antioxidative status. The results showed that MDA in cerebrum (112.4%) and sciatic nerve (113.1%) significantly increased (P<0.05) on third week of AC treatment and at gait score of 2, and further changes of MDA were observed after 6, 9, 12 weeks and at gait score of 3, 4. While a decrease (P<0.05) in the activities of GSH, CAT, GPx and SOD after 6, 9, 12 weeks intoxication and at gait score of 2, 3, 4 were observed in cerebrum, spinal cord and sciatic nerve. Anti-ROS activities also decreased in all three nerve tissues after 3, 6, 9, 12 weeks intoxication and at gait score of 2, 3, 4. Thus, AC intoxication was associated with elevation of lipid peroxidation and reduction of antioxidative status, and the time-dependent changes of these indexes in Wistar rats nerve tissues occurred. Sciatic nerve was the main target tissue and MDA was most sensitive among all indexes. The changes of lipid peroxidation and antioxidative status might be related to the degradation of nerve fiber and served as one of mechanisms of toxic neuropathy induced by AC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Allylchlorid, reagent grade, 98%
Sigma-Aldrich
Allylchlorid, ReagentPlus®, 99%
SAFC
Allylchlorid