Merck
  • The effect of ytterbium fluoride and barium sulphate nanoparticles on the reactivity and strength of a glass-ionomer cement.
The effect of ytterbium fluoride and barium sulphate nanoparticles on the reactivity and strength of a glass-ionomer cement.
Dental materials : official publication of the Academy of Dental Materials (2005-12-21)
Leon H Prentice, Martin J Tyas, Michael F Burrow
ZUSAMMENFASSUNG
While clinical advantages of glass-ionomers include fluoride release and radiopacity, disadvantages include low strength, slow initial setting times and opacity. The addition of nanoparticles, in particular those containing fluoride and cross-linkable ions, may mitigate the disadvantages while further improving the advantages. This investigation evaluated the effects of the addition of ytterbium fluoride (YbF3) and barium sulphate (BaSO4) on the strength and reactivity of a commercial glass-ionomer cement. YbF3 and BaSO4 nanoparticles were incorporated into the powder component of Riva SC (SDI Ltd., Bayswater, Australia) at 1, 2, 5, 10, 15, and 25% by weight. Capsules were assembled at a powder:liquid ratio of 2.9:1, activated and mixed, and the resultant pastes evaluated for working time, initial setting time, 24-h surface hardness and 24-h compressive strength. Working and initial setting times were reduced with the addition of YbF3. Addition of BaSO4 at low concentrations reduced working and initial setting times, but further addition delayed the setting reaction. Compressive strength decreased with the addition of either YbF3 or BaSO4, while surface hardness was slightly but insignificantly higher at 1-2% nanoparticles and then decreased with increasing nanoparticle concentrations. Nanoparticles modified the setting characteristics, strength and surface hardness of a commercial glass-ionomer cement, and may be useful for refining the handling characteristics of these materials. Further improvements in powder blending may result in more significant improvements in mechanical properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ytterbium(III)-fluorid, powder, 99.98% trace metals basis