Merck
Synthesis and characterization of new fluorene-based singlet oxygen sensitizers.
Journal of fluorescence (2005-12-21)
Kevin D Belfield, Claudia C Corredor, Alma R Morales, Marie A Dessources, Florencio E Hernandez
ZUSAMMENFASSUNG
The synthesis, photophysical characterization, and determination of singlet oxygen quantum yields (Phi(Delta)) for a class of fluorene derivatives with potential application in two-photon photodynamic therapy (PDT) is reported. It has been demonstrated that these compounds possess the ability to generate singlet oxygen (1O2) upon excitation. A photochemical method, using 1,3-diphenylisobenzofuran (DPBF) as 1O2 chemical quencher, was employed to determine the singlet oxygen quantum yields (Phi(Delta)) of the fluorene-based photosensitizers in ethanol. Phi(Delta) values ranged from 0.35 to 0.75. These derivatives may have potential application as two-photon photosensitizers when pumped via two-photon excitation in the near-IR spectral region.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Diphenyl-isobenzofuran, 97%