- [Spectrum studies and application of the reaction system phenanthroline-EBT-H2O2].
[Spectrum studies and application of the reaction system phenanthroline-EBT-H2O2].
Guang pu xue yu guang pu fen xi = Guang pu (2005-12-29)
Yu-you Zhou, Feng Gao, Shi-ke She, Qin Lu, Lun Wang
PMID16379299
ZUSAMMENFASSUNG
A kinetic spectrophotometric method for the determination of trace phenanthroline was proposed. The method is based on the inhibition of phenanthroline to the discoloring reaction of EBT oxidized by hydrogen peroxide in the dilute acetic acid solution. At the same time, the kinetic parameters were studied in detail. Under optimum experimental conditions, the linear range is 0.003-0.08 mg x L(-1), and the detection limit is 4.0 microg x L(-1). The method is simple, rapid, and easy to operate. The method was applied to the determination of trace phenanthroline in water samples with satisfactory results.