Merck
Removal of triazine herbicides from freshwater systems using photosynthetic microorganisms.
Environmental pollution (Barking, Essex : 1987) (2006-02-21)
O González-Barreiro, C Rioboo, C Herrero, A Cid
ZUSAMMENFASSUNG
The uptake of the triazine herbicides, atrazine and terbutryn, was determined for two freshwater photosynthetic microorganisms, the green microalga Chlorella vulgaris and the cyanobacterium Synechococcus elongatus. An extremely rapid uptake of both pesticides was recorded, although uptake rate was lower for the cyanobacterium, mainly for atrazine. Other parameters related to the herbicide bioconcentration capacity of these microorganisms were also studied. Growth rate, biomass, and cell viability in cultures containing herbicide were clearly affected by herbicide uptake. Herbicide toxicity and microalgae sensitivity were used to determine the effectiveness of the bioconcentration process and the stability of herbicide removal. C. vulgaris showed higher bioconcentration capability for these two triazine herbicides than S. elongatus, especially with regard to terbutryn. This study supports the usefulness of such microorganisms, as a bioremediation technique in freshwater systems polluted with triazine herbicides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Terbutrin, PESTANAL®, analytical standard