Merck
Arylamine-induced hemolytic anemia: electron spin resonance spectrometry studies.
Advances in experimental medicine and biology (1991-01-01)
T P Bradshaw, D C McMillan, R K Crouch, D J Jollow
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Phenylhydroxylamin, ≥95.0%