Merck
Anti-Plasmodium activity of imidazole-dioxolane compounds.
Bioorganic & medicinal chemistry letters (2006-02-24)
Jason Z Vlahakis, Robert T Kinobe, Kanji Nakatsu, Walter A Szarek, Ian E Crandall
ZUSAMMENFASSUNG
A series of imidazole-dioxolane compounds, which we hypothesize should bind to heme and thus interfere with heme catabolism in the parasite, were assayed for inhibitory activity in Plasmodium falciparum cultures and the results were compared to those obtained with Chinese hamster ovary (CHO) cells. The majority of the compounds displayed a similar ratio of inhibitory activity in the two culture systems; however, a number of the compounds tested showed promising anti-Plasmodium activity. The mechanism of action of these compounds remains unclear, however their inability to act synergistically with chloroquine suggests that, if they are inhibiting heme detoxification, they do so in a manner that does not complement the action of chloroquine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Dioxolan, ReagentPlus®, contains ~75 ppm BHT as inhibitor, 99%
Sigma-Aldrich
1,3-Dioxolan, anhydrous, contains ~75 ppm BHT as inhibitor, 99.8%