Merck
  • Pre-treatment of high oil and grease pet food industrial wastewaters using immobilized lipase hydrolyzation.
Pre-treatment of high oil and grease pet food industrial wastewaters using immobilized lipase hydrolyzation.
Journal of hazardous materials (2006-03-01)
Jeganaesan Jeganathan, Amarjeet Bassi, George Nakhla
ZUSAMMENFASSUNG
Wastewaters generating from pet food industries contain high concentration of oil and grease (O&G), which is difficult to treat through conventional biological treatment systems. In this study, the hydrolysis of O&G originating from pet food industrial wastewater was evaluated. Candida rugosa lipase was immobilized in calcium alginate beads and applied in the hydrolysis experiment. Results showed that approximately 50% of the O&G was hydrolyzed due to the enzyme activity. A significant increment in COD and VFA production was also observed. The immobilized lipase activity was confirmed with p-nitrophenyl palmitate (pNPP) before and after O&G hydrolysis. During the 3-day experiment, approximately 65% of the beads were recovered and after the hydrolysis, approximately 70% of the enzyme activity remained in the beads. This study shows the potential of immobilized lipase as a pre-treatment step in biological treatment of pet food manufacturing wastewater.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Nitrophenylpalmitat, lipase substrate