Merck
  • A simple and sensitive analytical method for the determination of antimony in environmental and biological samples.
A simple and sensitive analytical method for the determination of antimony in environmental and biological samples.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-03-04)
Kishore K Tiwari, Girdhar L Mundhara, Manish K Rai, Vinay K Gupta
ZUSAMMENFASSUNG
The results of a study and application of leucocrystal violet for the determination of antimony in parts per million levels is described here. The proposed method is based on the reaction of antimony(III) with acidified potassium iodate to liberate iodine. The liberated iodine selectively oxidizes leucocrystal violet to crystal violet dye. The formed dye shows maximum absorbance at 590 nm. The color system obeys Beer's law in the concentration range from 0.4 - 3.6 microg antimony per 25 ml of final solution. The molar absorptivity and Sandell's sensitivity were found to be 7.32 x 10(5) l mol(-1) cm(-1) and 0.0016 microg cm(-2), respectively. All variables were studied in order to optimize the reaction. The proposed method is satisfactorily applicable for the analysis of antimony in various environmental and biological samples. The method is simple, highly sensitive, accurate and reliable.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumiodat, ACS reagent, 99.5%
Sigma-Aldrich
Kaliumiodat, reagent grade, ≥98%
Sigma-Aldrich
Kaliumiodat, 99.995% trace metals basis
Supelco
Kaliumiodat, reference material for titrimetry, certified by BAM, >99.5%