Merck
  • Determination of chlormequat and mepiquat in foods by liquid chromatography/mass spectrometry or liquid chromatography/tandem mass spectrometry: interlaboratory study.
Determination of chlormequat and mepiquat in foods by liquid chromatography/mass spectrometry or liquid chromatography/tandem mass spectrometry: interlaboratory study.
Journal of AOAC International (2006-03-11)
Lutz Alder, James R Startin
ZUSAMMENFASSUNG
Interlaboratory validation studies have been performed on 2 methods for the determination of chlormequat (CLQ) and mepiquat (MPQ). Both methods used identical extraction procedures and stable isotope internal standardization but differed in the use of liquid chromatography/mass spectrometry (LC/MS) or LC/tandem mass spectrometry (LC/MS/MS) for the determination, the amount of internal standard used, and the expected limit of detection. After addition of deuterated internal standards, CLQ and MPQ were extracted with methanol-water and determined by LC//MS or LC/MS/MS with positive electrospray ionization. Eight European laboratories participated in the LC/MS method study, analyzing mushroom, pear, wheat flour, and fruit puree with residues of CLQ in the range 0.040-1.19 mg/kg and of MPQ in the range 0.041-0.39 mg/kg. For CLQ, the Horwitz ratio (HoRat) values for individual test materials/levels were in the range 0.85-1.13 with a mean of 1.00, showing good method performance. For MPQ, the Ho values for mushroom, pear (both levels), and wheat flour were in the range 0.83-0.94, again indicating good method performance. For the determination of MPQ in infant food (fruit puree) at 0.041 mg/kg, the Ho was 1.7 when a value of 0 reported by one participant was excluded. In the LC/MS/MS study, in which 11 laboratories participated, a separate sample set was analyzed with residues of CLQ in the range 0.007-1.03 mg/kg and of MPQ in the range 0.008-0.72 mg/kg. Ho values for CLQ were in the range 0.27-1.36 and for MPQ in the range 0.51-2.10, all corresponding to acceptable method performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(2-Chlorethyl)trimethylammoniumchlorid, 98%
Sigma-Aldrich
2-Chlorethyltrimethylammoniumchlorid, BioReagent, suitable for plant cell culture
Supelco
Mepiquatchlorid, PESTANAL®, analytical standard
Supelco
Chlormequatchlorid, PESTANAL®, analytical standard