Merck
  • Reduction of phosphoinositides and diacylglycerol levels in repeatedly dibutyltin-dilaurate-treated rat brain.
Reduction of phosphoinositides and diacylglycerol levels in repeatedly dibutyltin-dilaurate-treated rat brain.
Toxicology letters (1991-08-01)
A Subramoniam, R Husain, P K Seth
ZUSAMMENFASSUNG
Oral administration of a single dose of dibutyltin dilaurate (DBTL, 80 mg/kg body wt.), 2 or 24 h after treatment, caused no significant change in the levels of diacylglycerol and phosphoinositides in rat cerebrum (forebrain), whereas daily administration of DBTL (40 or 80 mg/kg body wt.) for 3 days (24 h after the final treatment) decreased the levels of diacylglycerol and phosphoinositides (phosphatidylinositol, phosphatidylinositol 4-phosphate and phosphatidylinositol 4,5-bisphosphate) in a concentration-dependent manner without influencing the levels of phosphatidylcholine in rat cerebrum. These studies indicate impairment of the phosphoinositide messenger system in rat cerebrum following repeated exposure to DBTL.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dibutylzinndilaurat, 95%