Merck
  • An efficient stereoselective dihydroxylation of glycals using a bimetallic system, RuCl3/CeCl3/NaIO4.
An efficient stereoselective dihydroxylation of glycals using a bimetallic system, RuCl3/CeCl3/NaIO4.
The Journal of organic chemistry (2006-03-25)
Pallavi Tiwari, Anup Kumar Misra
ZUSAMMENFASSUNG
A catalytic dihydroxylation reaction on glycals has been developed using a bimetallic oxidizing system to furnish sugar 1,2-diols in a highly stereoselective manner.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ruthenium(III)-chlorid, Anhydrous, solid, Ru content 45-55%