Merck
  • In search of excited-state proton transfer in the lumichrome dimer in the solid state: theoretical and experimental approach.
In search of excited-state proton transfer in the lumichrome dimer in the solid state: theoretical and experimental approach.
The journal of physical chemistry. A (2006-04-08)
Ewa Sikorska, Igor Khmelinskii, Maciej Kubicki, Wiesław Prukała, Marcin Hoffmann, Isabel F Machado, Luis F V Ferreira, Jerzy Karolczak, David R Worrall, Alina Krawczyk, Małgorzata Insińska-Rak, Marek Sikorski
ZUSAMMENFASSUNG
Quantum chemical density functional theory (DFT) calculations and spectral data were employed to investigate the possibility of the excited-state double proton transfer (ESDPT) in lumichrome crystals. The calculations in a lumichrome dimer predict a transfer of a proton in the first excited state, leading to a cation-anion pair. The presently reported X-ray structure of 1,3-dimethyllumichrome and its complex solid-state luminescence indicate that also in this molecule intermolecular hydrogen bonds might be involved in the photophysics. The long-wavelength emission in lumichrome crystals peaked at 530 nm is attributed to excited-state proton transfer, whereas a wider emission band in methylated lumichrome derivatives peaked at 560 nm is attributed to ions formed upon photoexcitation of the crystals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lumichrom