Merck
  • A quinhydrone-type 2 ratio 1 acceptor-donor charge transfer complex obtained via a solvent-free reaction.
A quinhydrone-type 2 ratio 1 acceptor-donor charge transfer complex obtained via a solvent-free reaction.
Chemical communications (Cambridge, England) (2006-04-13)
Marcel Bouvet, Bernard Malézieux, Patrick Herson
ZUSAMMENFASSUNG
A 2 : 1 2-methoxybenzoquinone (BQ(OMe))-hydroquinone (H2Q) complex (5), obtained as single crystals by a solvent-free reaction, combines dipolar and pi-pi interactions, as well as hydrogen bonding.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chinhydron, 97%