Merck
  • (99m)Tc complexes with activated ester functions; ligands comprising a 3,4-diamino-benzoate backbone.
(99m)Tc complexes with activated ester functions; ligands comprising a 3,4-diamino-benzoate backbone.
Nuclear medicine and biology (2006-04-25)
Osmar Calderon Sanchez, Ashour Mohammed, Claudia Bauer, Markus Wolf, Björn Wängler, Walter Mier, Uwe Haberkorn, Raoul Mocelo, Michael Eisenhut
ZUSAMMENFASSUNG
Preformed (99m)Tc chelates with an activated ester function are useful for the gentle labeling of proteins (precomplexation route). In this context, new heterobifunctional ligands derived from 2,3,5,6-tetrafluorophenyl (TFP) 3,4-diamino-benzoates (OC1, OC3, OC4) were synthesized. Their corresponding (99m)Tc-complexation and protein-conjugation characteristics were elucidated and compared with the results previously reported using 2,3,5,6-tetrafluorophenyl N-(S-benzoylthioacetyl)glycylglycyl-p-aminobenzoate (OC2). The reaction temperatures and the reaction time markedly influenced complexation yields. Compared with OC2, the (99m)Tc-chelate formation with OC1 and OC4 was more effective, showing radiochemical yields of 60% and 70% within 20 min, respectively. Owing to steric hindrance, the complexation of OC3, however, did not exceed 10%. No-carrier-added (99m)Tc complexes were conjugated at pH 10 with the anti-EGF-receptor monoclonal antibody MAb425, resulting in labeling yields of 14% for (99m)Tc-OC1 and 7% for (99m)Tc-OC4 after incubating for 20 min at 30 degrees C. Increasing the temperature to 40 degrees C improved these results by 14% and 3%, respectively. As compared with (99m)Tc-OC2, which provides the chelating substituent at the 4-phenyl position only, the application of 3,4-phenyl substituents proved less appropriate for protein conjugation. However, the 3,4-diaminobenzoate backbone may be attractive for an alternative design of novel (99m)Tc N2S2 or N3S complexes, because they show excellent complexation characteristics.