- Low background FRET-substrates for lipases and esterases suitable for high-throughput screening under basic (pH 11) conditions.
Low background FRET-substrates for lipases and esterases suitable for high-throughput screening under basic (pH 11) conditions.
Organic & biomolecular chemistry (2006-04-25)
Yongzheng Yang, Peter Babiak, Jean-Louis Reymond
PMID16633567
ZUSAMMENFASSUNG
FRET-based fluorogenic substrates for lipases and esterases were prepared in four steps from commercially available building blocks. The substrates are pyrenebutyric acid monoesters of aliphatic 1,2-diols bearing a dinitrophenylamino group as a quencher. The most enzyme-reactive substrate is ester 2a. The substrates do not show any measurable background reaction in the absence of enzyme even at pH 11, but react fast and specifically with lipases and esterases. These substrates offer an unprecedented and practical solution to the long-standing problem of a simple yet efficient high-throughput screening tool for lipase activities under basic conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.