Merck
  • [Reactions of 1-(X-benzoyl)-4-R-thiosemicarbazide with chloroacetone and omega-bromoacetophenone. II. 4-Phenyl- and 4-(p-tolyl)-thiosemicarbazide of p-nitrobenzoic acid].
[Reactions of 1-(X-benzoyl)-4-R-thiosemicarbazide with chloroacetone and omega-bromoacetophenone. II. 4-Phenyl- and 4-(p-tolyl)-thiosemicarbazide of p-nitrobenzoic acid].
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina (1990-01-01)
E Bielak, S Biliński
ZUSAMMENFASSUNG
The reactions of 4-phenyl- and 4-(p-tolyl)-thiosemicarbazide of p-nitrobenzoic acid (Ic, d) with chloroacetone and omega-bromoacetophenone were investigated in: 1) methanolic medium (method D); 2) methanolic medium in the presence of anhydrous CH3COONa (method E); 3) methanolic medium in the presence of N(C2H5)3 (method F). The properties of compounds IIe-h and IIIe-h were determined under the conditions of basic and acidic hydrolysis. The results of UV and IR spectroscopic measurements were reported.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloraceton, 95%
Sigma-Aldrich
Chloraceton, Wacker Chemie AG, ≥96.0% (GC)