Merck
  • Further evidence of an inverted region in proton transfer within the benzophenone/substituted aniline contact radical ion pairs; importance of vibrational reorganization energy.
Further evidence of an inverted region in proton transfer within the benzophenone/substituted aniline contact radical ion pairs; importance of vibrational reorganization energy.
The journal of physical chemistry. A (2006-05-19)
Libby R Heeb, Kevin S Peters
ZUSAMMENFASSUNG
The dynamics of proton transfer within the triplet contact radical ion pair of a variety of substituted benzophenones with N,N-diethylaniline, N,N-dimethyl-p-toluinide, and N,N-diallylaniline are examined in solvents of varying polarity. The correlation of the rate constants with driving force reveal both a normal region and an inverted region providing support for the nonadiabatic nature of proton transfer within these systems. The reorganization of both the solvent and the molecular framework are central in governing the overall reaction dynamics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4,N,N-Trimethylanilin, 99%
Sigma-Aldrich
2,6-Diethylanilin, 98%
Sigma-Aldrich
4,N,N-Trimethylanilin, ≥98.5% (GC)
Sigma-Aldrich
4,N,N-Trimethylanilin, purum, ≥98.0% (GC)