Merck
  • Contribution of spinal glutamatergic receptors to the antinociception caused by agmatine in mice.
Contribution of spinal glutamatergic receptors to the antinociception caused by agmatine in mice.
Brain research (2006-06-13)
Vinicius M Gadotti, Daiane Tibola, Ana Flavia Paszcuk, Ana Lúcia S Rodrigues, João B Calixto, Adair R S Santos
ZUSAMMENFASSUNG
This study was designed to evaluate the role of spinal glutamatergic receptors in the antinociception elicited by agmatine in mice. Intraperitoneal (i.p.) administration of agmatine (1.0-100.0 mg/kg) dose dependently inhibited the nociceptive response induced by intrathecal (i.t.) injection of glutamate, N-methy-D-aspartate (NMDA) and (+/-)-1-aminocyclopentane-trans-1,3-dicarboxylic acid (trans-ACPD), with mean ID(50) values of 16.7, 6.8 and 27.0 mg/kg, respectively. However, agmatine completely failed to affect the nociception induced by alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) or kainic acid (kainate). Agmatine injected by i.t. route (10-100 microg/site) also produced dose-related inhibition of NMDA- and trans-ACPD-induced biting response with mean ID(50) values of 29.6 and 36.0 mug/site, respectively. The nitric oxide synthase inhibitor N(omega)-nitro-L-arginine (l-NOARG) (75.0 mg/kg, i.p.) also consistently inhibited glutamate-, NMDA- and trans-ACPD-induced nociception (41 +/- 13, 100 and 83 +/- 6%, of inhibition, respectively) but had no effect on the same response caused by AMPA and kainate agonists. The selective NMDA receptor antagonist (5S,10R)-(+)-5-Methyl-10,11-dihydro-5H-dibenzo[a,d] (MK-801) at a low dose (0.05 mg/kg, i.p.) inhibited the nociceptive response caused by both glutamate and NMDA agonists (inhibitions of 35 +/- 1 and 72 +/- 2%, respectively). At a high dose, MK-801 (0.5 mg/kg, i.p.) significantly inhibited the biting response induced by i.t. administration of all the glutamatergic agonists tested: glutamate, AMPA, NMDA, kainate and trans-ACPD, with inhibitions of 49 +/- 8, 84 +/- 16, 84 +/- 3, 76 +/- 8 and 97 +/- 2%, respectively. Together, these results provide experimental evidence indicating that agmatine given systemically and spinally produce marked antinociception at spinal sites in mice. Furthermore, an interaction with glutamate receptors, namely NMDA and trans-ACPD, metabotropic and NMDA-ionotropic origin, by a mechanism similar to that of nitric oxide (NO) inhibitors, seems to account for the agmatine antinociceptive action.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
trans-(±)-ACPD monohydrate, solid, ≥97% (NMR)