Merck
Hydrothermal preparation and luminescence of LaF3:Eu3+ nanoparticles.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-07-04)
Jian-Xin Meng, Mao-Feng Zhang, Ying-Liang Liu, Shi-Qing Man
ZUSAMMENFASSUNG
LaF3:Eu3+ nanoparticles were prepared by a simple hydrothermal process at low temperature and characterized by X-ray diffraction (XRD), transmission electron microscopy (TEM) and fluorescence spectrum. Well-dispersed nanoparticles with an average size of 30 nm and a hexagonal shape were obtained. The influences of reaction temperature and time on the preparation and luminescence of LaF3:Eu3+ nanoparticles were investigated. Luminescent quenching occurred at a much higher concentration ( approximately 25mol%) and stronger luminescent intensity than in bulk LaF3:Eu3+. Fluorescence intensity of the LaF3:Eu3+ nanoparticles varied remarkably with calcination temperatures. It was found that samples without any further calcinations can emit quite strong fluorescence.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lanthan(III)-fluorid, powder, 99.99% trace metals basis