Merck
  • Spectrophotometric determination of trazodone, amineptine and amitriptyline hydrochlorides through ion-pair formation with molybdenum and thiocyanate.
Spectrophotometric determination of trazodone, amineptine and amitriptyline hydrochlorides through ion-pair formation with molybdenum and thiocyanate.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-07-11)
Gehad G Mohamed, F A Nour El-Dien, S M Khalil, Nehad A Mohamed
ZUSAMMENFASSUNG
Extraction spectrophotometric method has been developed for the determination of tricyclic drugs such as trazodone (TZH), amineptine (APH) and amitriptyline (ATPH) hydrochlorides in pure form and in the dosage forms coming from different Egyptian markets. The method based on the formation of ion-pairs between these drugs under investigation and inorganic complex of Mo(V)-thiocyanate followed by its extraction with methylene chloride. The optimum conditions for the ion-pairs formation are established. The method permits the determination of TZH, APH and ATPH over the concentration range of 2-28, 2-32 and 1-30 microg ml(-1), respectively. The Sandell sensitivity (S) is found to be 0.105, 0.138 and 0.118 g cm(-2) for TZH, APH and ATPH, respectively. The SD is found to be 0.16-0.377, 0.12-0.259 and 0.091-0.286 and the R.S.D. are 0.14-0.55, 0.12-0.399 and 0.095-0.485 for TZH, APH and ATPH, respectively. The method is applicable for the assay of the investigated drugs in different dosage forms and the results are in good agreement with those obtained by the official method.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniummolybdat Tetrahydrat, ACS reagent, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammoniumthiocyanat, ACS reagent, ≥97.5%
Sigma-Aldrich
Ammoniummolybdat Tetrahydrat, ACS reagent, 99.98% trace metals basis
Sigma-Aldrich
Ammoniummolybdat Tetrahydrat, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammoniummolybdat, 99.98% trace metals basis
Sigma-Aldrich
Ammoniummolybdat Tetrahydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammoniumthiocyanat, crystals
Sigma-Aldrich
Ammoniummolybdat, meets USP testing specifications
Supelco
Ammoniumthiocyanat, 99.99% trace metals basis