Merck
  • Characterization of amino acids in silk sericin protein from Gonometa rufobrunnae by MEKC with phenyl isothiocyanate derivatization.
Characterization of amino acids in silk sericin protein from Gonometa rufobrunnae by MEKC with phenyl isothiocyanate derivatization.
Journal of separation science (2006-07-13)
Simiso Dube, Mkhululi T Khumalo, Nelson Torto, Jacob A Nyati
ZUSAMMENFASSUNG
An MEKC method was developed for the separation and characterization of phenyl-isothiocyanate (PITC)-labeled amino acids derived from Gonometa rufobrunnae silkworm after microdialysis sample cleanup. The influence of the buffer and SDS concentration on the resolution of the amino acids was investigated. A buffer system consisting of 25 mM phosphate, 10 mM borate buffer at pH 9.00, and 70 mM SDS showed the best results, with 13 PITC-amino acid derivatives being resolved out of 15 possible amino acids that were under study. Microdialysis sampling demonstrated its efficiency as a sample cleanup technique. Sericin protein from G. rufobrunnae was found to be characterized by at least 11 positively identified amino acids. These included His, Tyr, Ser, Ala, Phe, Lys, Gly, Arg, Cys, Glu, and Asp. Leu/Met and Val/Thr were coeluting pairs and hence could not be positively confirmed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenylisothiocyanat, reagent grade, 98%
Sigma-Aldrich
Phenylisothiocyanat, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Phenylisothiocyanat, 99%, for protein sequencing
Sigma-Aldrich
Phenylisothiocyanat, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid