Merck
Tuning the structure of SDS micelles by substituted anilinium ions.
The journal of physical chemistry. B (2006-07-21)
Gunjan Garg, P A Hassan, V K Aswal, S K Kulshreshtha
ZUSAMMENFASSUNG
The growth behavior of aggregates formed in aqueous solutions of the anionic surfactant sodium dodecyl sulfate (SDS) in the presence of the cationic hydrophobic salts o-toluidine hydrochloride (OTHC) and m-toluidine hydrochloride (MTHC) has been studied by dynamic light scattering (DLS) and small-angle neutron scattering (SANS) techniques. DLS studies indicate a progressive growth of SDS micelles with addition of less than equimolar concentrations of hydrophobic salts. A prolate ellipsoidal model is used to analyze the DLS data, which is further supported by SANS measurements. We explain the propensity for the strong growth of micelles in the presence of OTHC and MTHC by the high charge neutralization provided by these salts as the aromatic counterions are adsorbed on the surface of the micelles. When the substitution is at the meta position, i.e., for MTHC, micellar growth is favored at lower salt concentrations than for OTHC. The variation in growth behavior is explained in terms of the difference in the chemical environments of the substituents at the ortho and meta positions. Micellar parameters obtained from SANS data at elevated temperature also support enhanced growth of micelles in the presence of MTHC as compared to OTHC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
m-Toluidin, 99%