Merck
A microchip sensor for calcium determination.
Analytical and bioanalytical chemistry (2006-10-05)
P Caglar, S A Tuncel, N Malcik, J P Landers, J P Ferrance
ZUSAMMENFASSUNG
A newly designed glass-PDMS microchip-based sensor for use in the determination of Ca(2+) ions has been developed, utilizing reflectance measurements from arsenazo III (1,8-dihydroxynaphthalene-3,6-disulfonic acid-2,7-bis[(azo-2)-phenyl arsenic acid]) immobilized on the surface of polymer beads. The beads, produced from cross-linked poly(p-chloromethylstyrene) (PCMS), were covalently modified with polyethylenimine (PEI) to which the Arsenazo III could be adsorbed. The maximum amount of Arsenazo III which could be immobilized onto the PEI-attached PCMS beads was found to be 373.71 mg g(-1) polymer at pH 1. Once fabricated, the beads were utilized at the detection point of the microfluidic sensor device with a fiber optic assembly for reflectance measurements. Samples were mobilized past the detection point in the sensor where they interact with the immobilized dye. The sensor could be regenerated and re-used by rinsing with HCl solution. The pH, voltage, linear range, and the effect of interfering ions were evaluated for Ca(2+) determination using this microchip sensor. At the optimum potential, 0.8 kV, and pH 9.0, the linear range of the microchip sensor was 3.57 x 10(-5) - 5.71 x 10(-4) M Ca(2+), with a limit of detection (LOD) of 2.68 x 10(-5) M. The microchip biosensor was then applied for clinical analysis of calcium ions in serum with good results.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Arsenazo III, calcium-sensitive dye