Merck
  • Hypolipidaemic and antiplatelet activity of phenoxyacetic acid derivatives related to alpha-asarone.
Hypolipidaemic and antiplatelet activity of phenoxyacetic acid derivatives related to alpha-asarone.
The Journal of pharmacy and pharmacology (2006-10-13)
Ricardo Pérez-Pastén, Rosa Virginia García, Leticia Garduño, Elba Reyes, Fernando Labarrios, Joaquín Tamariz, Germán Chamorro
ZUSAMMENFASSUNG
The phenoxyacetic acid derivatives 1-6 [2-methoxy-4-(2-propenyl)phenoxyacetic acid (1); 2-methoxy-5-nitro-4-(2-propenyl)phenoxyacetic acid (2); methyl 2-methoxy-4-(2-propenyl)phenoxyacetate (3); ethyl 2-methoxy-4-(2-propenyl)phenoxyacetate (4); methyl 2-methoxy-5-nitro-4-(2-propenyl)phenoxyacetate (5); ethyl 2-methoxy-5-nitro-4-(2-propenyl)phenoxyacetate (6)] related to alpha-asarone have been reported previously as hypolipidaemic agents in diet-induced hyperlipidaemic mice. We have aimed to expand the pharmacological profile of these derivatives by investigating their hypolipidaemic activity in rats and mice under different experimental conditions. The antiplatelet activity was tested also in-vitro from blood derived from consenting healthy volunteers. In normolipidaemic rats, compounds 2, 3 and 5 at oral doses of 40 and 80 mg kg(-1) significantly decreased total cholesterol and LDL-cholesterol levels. Moreover, analogues 3 and 5 administered to hypercholesterolaemic rats at the same doses for seven days also produced a reduction in the content of these same lipoproteins. In neither case were the high-density lipoprotein cholesterol and triglyceride concentrations affected. However, practically all tested compounds were found to be hypocholesterolaemic agents, and were shown to effectively lower low-density lipoprotein cholesterol and triglyceride levels in Triton-induced hyperlipidaemic mice at oral doses of 50 and 100 mg kg(-1). In all tests, all animals appeared to be healthy throughout the experimental period in their therapeutic ranges. Triton-induced hypercholesterolaemic mice appeared to be a desirable model for this class of hypolipidaemic drugs. On the other hand, compounds 1, 2, 4 and 5 significantly inhibited ADP-induced aggregation in-vitro. These findings indicated that all of these compounds appeared to be promising for the treatment of human hyperlipidaemia and thrombotic diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenoxyessigsäure, 98%
Supelco
Phenoxyessigsäure, PESTANAL®, analytical standard