Merck
Coimmobilization of a redox enzyme and a cofactor regeneration system.
Chemical communications (Cambridge, England) (2006-10-19)
Lorena Betancor, Cécile Berne, Heather R Luckarift, Jim C Spain
ZUSAMMENFASSUNG
The coimmobilization of nitrobenzene nitroreductase and glucose-6-phosphate dehydrogenase in silica particles enables the continuous conversion of nitrobenzene to hydroxylaminobenzene with NADPH recycling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Phenylhydroxylamin, ≥95.0%