Merck
  • Increased Rho activation and PKC-mediated smooth muscle contractility in the absence of caveolin-1.
Increased Rho activation and PKC-mediated smooth muscle contractility in the absence of caveolin-1.
American journal of physiology. Cell physiology (2006-11-15)
Yulia Shakirova, Johan Bonnevier, Sebastian Albinsson, Mikael Adner, Bengt Rippe, Jonas Broman, Anders Arner, Karl Swärd
ZUSAMMENFASSUNG
Caveolae are omega-shaped membrane invaginations that are abundant in smooth muscle cells. Since many receptors and signaling proteins co-localize with caveolae, these have been proposed to integrate important signaling pathways. The aim of this study was to test whether RhoA/Rho-kinase and protein kinase C (PKC)-mediated Ca(2+) sensitization depends on caveolae using caveolin (Cav)-1-deficient (KO) and wild-type (WT) mice. In WT smooth muscle, caveolae were detected and Cav-1, -2 and -3 proteins were expressed. Relative mRNA expression levels were approximately 15:1:1 for Cav-1, -2, and -3, respectively. Caveolae were absent in KO and reduced levels of Cav-2 and Cav-3 proteins were seen. In intact ileum longitudinal muscle, no differences in the responses to 5-HT or the muscarinic agonist carbachol were found, whereas contraction elicited by endothelin-1 was reduced. Rho activation by GTPgammaS was increased in KO compared with WT as shown using a pull-down assay. Following alpha-toxin permeabilization, no difference in Ca(2+) sensitivity or in Ca(2+) sensitization was detected. In KO femoral arteries, phorbol 12,13-dibutyrate (PDBu)-induced and PKC-mediated contraction was increased. This was associated with increased alpha(1)-adrenergic contraction. Following inhibition of PKC, alpha(1)-adrenergic contraction was normalized. PDBu-induced Ca(2+) sensitization was not increased in permeabilized femoral arteries. In conclusion, Rho activation, but not Ca(2+) sensitization, depends on caveolae in the ileum. Moreover, PKC driven arterial contraction is increased in the absence of caveolin-1. This depends on an intact plasma membrane and is not associated with altered Ca(2+) sensitivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cirazoline hydrochloride, solid